Learn More
Description
Includes
MTT reagent; Detergent Reagent
Tetrazolium compound MTT changes to from yellow to purple
MTT is added directly to the culture medium and is reduced by metabolically active cells to insoluble purple formazan dye crystals
The absorbance of the sample is read directly in the wells at an
Specifications
Specifications
| Quantity | 5000 Tests |
| Product Type | MTT Cell Proliferation/Viability Assay |
| Content And Storage | The product requires storage at 2 - 8° C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date. |
| Product Line | R&D Systems |
| For Use With (Application) | Minimizes steps to assay for proliferation rate in cultured cells |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
