Description
The activated NEDD8 is transferred to UbcH12, and this E2-S-Ub thiolester complex can be used for the conjugation of NEDD8 to protein substrates in the presence of the appropriate E3 enzymes (not supplied)
Specifications
Specifications
| Content And Storage | Store the unopened product at -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 1 Kit |