Learn More
Description
Includes
1mL 10X NEDD8 Reaction Buffer; 100μL 10X Mg-ATP; 1mL 50X E1 Stop Buffer; 1mL 5X Non-reducing gel loading buffer
NEDD8 is expressed in striated muscle, and via its C-terminal Gly, forms covalent bonds with cullin family proteins. This conjugation generates ubiquitin ligase activity that regulates cell cycle proteins.
Buffer can be used to supplement NEDD8 conjugation kit (Mfr. No. K-800)
Includes recommended protocol for SDS-PAGE/Western Blot
Specifications
Specifications
| Quantity | 1 kit |
| Storage Requirements | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
