Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NK 252
GREENER_CHOICESDP

Catalog No. 529350 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
529350 50 mg
529310 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 529350 Supplier Tocris Bioscience™ Supplier No. 5293/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nrf2 activator

NK 252 is an Nrf2 activator; interacts with the Nrf2-binding site of Keap1. Downregulates the expression of fibrogenic genes and increases antioxidant effects in NASH rat models of liver disease.

Specifications

Chemical Name or Material NK 252
CAS 1414963-82-8
Synonym N-[5-(2-Furanyl)-1,3,4-oxadiazol-2-yl]-N'-(2-pyridinylmethyl)urea
Quantity 50 mg
Solubility Soluble to 100mM in 1eq. HCl and to 100mM in DMSO
Molecular Formula C13H11N5O3
Formula Weight 285.26
Physical Form Powder
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.