Learn More
Tocris Bioscience™ OICR 9429

Description
To request the negative control for OICR 9429, please fill out the OICR 0547 request form on the SGC website.
Specifications
Specifications
| Chemical Name or Material | OICR 9429 |
| Synonym | N-[2-(4-Methylpiperazin-1-yl)-5-[3-(morpholin-4-ylmethyl)phenyl]phenyl]-6-oxo-4-(trifluoromethyl)-1,6-dihydropyridine-3-carboxamide |
| Quantity | 10 mg |
| Solubility | Soluble to 50mM in DMSO |
| Target | Additional Lysine Methyltransferase Compounds |
| Molecular Formula | C29H32F3N5O3 |
| Formula Weight | 555.59 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.