Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PFI 4

Catalog No. 557610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
557610 10 mg
557650 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 557610 Supplier Tocris Bioscience™ Supplier No. 5576/10
This item is not returnable. View return policy

Potent and selective BRPF1 bromodomain inhibitor

PFI 4 is a potent and selective BRPF1B bromodomain inhibitor (IC50 = 172 nM). Exhibits >100-fold selectivity for BRPF1 over a panel of other bromodomains including BRPF2 (BRD1), BRPF3 and BRD4. Disrupts BRPF1 binding to histone H3.3. Cell permeable. Inhibits osteoclastogenesis.

Specifications

Chemical Name or Material PFI 4
CAS 900305-37-5
Synonym N-[2,3-Dihydro-1,3-dimethyl-2-oxo-6-(1-pyrrolidinyl)-1H-benzimidazol-5-yl]-2-methoxybenzamide
Quantity 10 mg
Solubility Soluble to 50mM in DMSO and to 10mM in ethanol
Molecular Formula C21H24N4O3
Formula Weight 380.44
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >99%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.