Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PFK 15
GREENER_CHOICE

Catalog No. 533910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
533910 10 mg
533950 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 533910 Supplier Tocris Bioscience™ Supplier No. 5339/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective PFKFB3 inhibitor

PFK 15 is an originally reported to be a selective PFKFB3 inhibitor (IC50 = 207 nM); a more recent report failed to demonstrate activity in a PFKFB3 kinase assay. Selective for PFKFB3 over a panel of 96 kinases. Reduces glucose uptake and induces apoptosis in H522 lung adenocarcinoma and Jurkat T cell leukemia cells. Suppresses glucose uptake and tumor growth, and inhibits metastasis of Lewis lung carcinoma cell xenografts in mice.

Specifications

Chemical Name or Material PFK 15
CAS 4382-63-2
Synonym 1-(4-Pyridinyl)-3-(2-quinolinyl)-2-propen-1-one
Quantity 10 mg
Solubility Soluble to 100mM in DMSO
Molecular Formula C17H12N2O
Formula Weight 260.29
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >99%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.