Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PR 619
GSA_VAGREENER_CHOICE

Catalog No. 448210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
448210 10 mg
448250 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 448210 Supplier Tocris Bioscience™ Supplier No. 4482/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Broad spectrum DUB inhibitor

PR 619 is a broad spectrum, reversible DUB inhibitor. Exhibits limited activity against other proteases. Induces accumulation of polyubiquitinated proteins, but has no direct inhibitory effect on the proteasome. Cytotoxic in HEK293T and colorectal cancer cells. Stabilizes microtubule network in oligodendroglial cells.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Chemical Name or Material PR 619
CAS 2645-32-1
Synonym 2,6-Diamino-3,5-dithiocyanopyridine
Quantity 10 mg
Solubility Soluble to 10mM in DMSO
Molecular Formula C7H5N5S2
Formula Weight 223.28
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.