Description
Proteinase K is one of the most active endopeptidases known. It rapidly inactivates endogenous nucleases including RNases and DNases. Permeabilization of the sample by Proteinase K creates pores in the cell membrane and facilitates access to the DNA by biotinylated dNTPS.
Specifications
Specifications
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 30 μL |
| Product Type | Proteinase K |
| Product Line | R&D Systems |