Learn More
R&D Systems™ Proteome Purify 12 Human Serum Protein Immunodepletion Resin
Click to view available options
Quantity:
20 Tests
40 Tests
Description
Includes
Immunodepletion Resin, Spin-X Filter Units
Depletes samples of the 12 high-abundance proteins comprising approximately 95% of proteins in human serum or plasma
Depletes samples of albumin, IgA, IgG and IgM, Fibrinogen, Transferrin, Haptoglobin and others
Removal allows visualization of proteins
Specifications
Specifications
Quantity | 40 Tests |
Product Type | Immunodepletion Resin |
Content And Storage | Store the unopened product at 2 - 8° C. Do not use past expiration date. |