Learn More
R&D Systems™ Proteome Purify 2 Mouse Serum Protein Immunodepletion Resin
Catalog No. p-4775711
Click to view available options
Quantity:
20 Tests
40 Tests
This item is not returnable. View return policy
This item is not returnable. View return policy
Provides an easy-to-use, high capacity system to reproducibly remove albumin and IgG from mouse serum or plasma
Analysis of low abundance proteins by even the most sensitive of methods for proteomics greatly benefits from the removal of high abundance proteins that may interfere with their detection.
  • Improves visualization of target proteins
  • Allows user to increase protein load or concentration
Specifications
Quantity 40 Tests
Product Type Immunodepletion Resin
Content And Storage Store the unopened product at 2 - 8° C. Do not use past expiration date.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.