Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ QAQ Dichloride

Catalog No. p-7082194
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Photoswitchable Nav, Kv and Cav channel blocker

QAQ dichloride is a photoswitchable Nav, Cav and Kv channel blocker. Blocks channels in the trans conformation. Switches conformation from cis to trans at 500 nm and trans to cis at 380 nm. Normally membrane impermeant, but enters nociceptive neurons via ion channels activated by noxious stimuli, particularly TRPV1. Exhibits optically reversible local anesthesia in rats.

Specifications

Chemical Name or Material QAQ dichloride
Synonym 2,2'-[1,2-Diazenediylbis(4,1-phenyleneimino)bis[N,N,N-triethyl-2-oxo-ethanaminium] dichloride
Quantity 10 mg
Solubility Soluble to 50mM in water
Target Non-selective Cav Blockers
Molecular Formula C28H44Cl2N6O2
Formula Weight 567.59
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.