Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Rabbit Protein Fraction II Protein Fraction
Click to view available options
Quantity:
5 mg
Description
Source: Rabbit erythrocytes,
Formulation: Supplied as a solution in HEPES.
Regulatory Status: RUO
Specifications
Specifications
Formulation | Supplied as a solution in HEPES. |
Name | Protein Fraction II |
Quantity | 5 mg |
Conjugate | Unconjugated |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction