Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
R&D Systems™ Recombinant Rat CCL2/JE/MCP-1 Protein
                    Click to view available options
                
                
                    
                            
                                Formulation:
                            
                        
                        
                            
                                Lyophilized from a 0.2μm filtered solution in PBS with Trehalose and with BSA as a carrier protein.
                            
                        
                            
                                Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
                            
                        
                    Description
The Recombinant Rat CCL2/JE/MCP-1 Protein is derived from NS0. The Recombinant Rat CCL2/JE/MCP-1 Protein has been validated for the following applications: Bioactivity.
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
                
            Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | 
| Gene ID (Entrez) | 24770 | 
| Molecular Weight (g/mol) | 14 kDa | 
| Name | CCL2/JE/MCP-1 | 
| Quantity | 25 μg | 
| Source | Mouse myeloma cell line,NS0-derived rat CCL2/JE/MCP-1 protein Gln24-Asn148 | 
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | 
| Conjugate | Unconjugated | 
| Recombinant | Recombinant | 
| Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            