Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Rat GM-CSF Protein

Catalog No. 518GM005CF
Click to view available options
Quantity:
25 μg
5 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Supplied as a 0.2μm filtered solution in PBS.
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Rat GM-CSF Protein is derived from E. coli. The Recombinant Rat GM-CSF Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Supplied as a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 116630
Molecular Weight (g/mol) 14.7 kDa
Name GM-CSF
Quantity 5 μg
Source E. coli-derived rat GM-CSF protein Ala1-Lys127,with and without an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after opening.
Gene Alias colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, CSF2, CSF-2, GMCSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, Molgramostim, molgramostin, sargramostim
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.