Learn More
Description
Recombinant Human YOD1 is a Ubiquitin--specific deconjugating enzyme. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human YOD1 concentration of 0.1-1 μM.
Specifications
Specifications
| Quantity | 50 μg |
| Species | Human |
| Conjugate | Unconjugated |
| Product Line | R&D Systems |
| Synonym | DKFZp451J1719, DUBA8, DUBA-8, DUBA8hsHIN7, EC 3.1.2, EC 3.4.19.12, HIN7, HIN-7, HIV-1-induced protease 7, HsHIN7, OTU domain-containing protein 2, OTUD2, OTUD2OTU domain containing 2, PRO0907, ubiquitin thioesterase OTU1, YOD1 OTU deubiquinating enzyme 1 homolog (yeast), YOD1 OTU deubiquinating enzyme 1 homolog (S. cerevisiae) |
| Content And Storage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
