Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse Reg3B Protein
Click to view available options
Quantity:
50 μg
Description
The Recombinant Mouse Reg3B Protein is derived from E. coli. The Recombinant Mouse Reg3B Protein has been validated for the following applications: Bioactivity.
Order Info
The product is shipped at ambient temperature. Upon receipt, store it immediately at the recommended storage temperatures.
Specifications
Specifications
Gene ID (Entrez) | 18489 |
Quantity | 50 μg |
Source | E. coli-derived mouse Reg3B protein Glu27-Gly175,with an N-terminal Met |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Gene Alias | PAP1, Reg3B |
Conjugate | Unconjugated |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction