Learn More
R&D Systems™ StemXVivo Mouse/Rat Osteogenic Supplement, 12.5 mL

Description
Despite the well-characterized factors and protocols used to differentiate mesenchymal stem/stromal cells (MSCs) into osteocytes, differentiation efficiencies can vary depending on the quality of the MSC starting population and the reagents used to expand and differentiate MSCs.
- Contains high quality differentiation factors to drive reproducible and efficient MSC osteogenesis
- Is defined to reduce unwanted experimental variability
- Has been developed and optimized using MSCs
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Product Type | Stem Cell Supplement Media |
| Quantity | 12.5 mL |
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.