Description
Includes
1mL 10X SUMO Reaction Buffer; 100μL 10X Mg-ATP; 1mL 50X E1 Stop Buffer; 1mL 5X Non-reducing gel loading buffer
This kit can supplement the SUMO conjugation kits (Mfr. Nos. K-712, K-715, K-720)
Includes recommended protocol for SDS-PAGE/Western Blot
Specifications
Specifications
| Storage Requirements | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |