Learn More
Description
Potent and selective GPR39 agonist (EC50 values are 0.4 and 0.8 nM for rat and human receptors respectively). Selective over a panel of kinases (IC50s > 10 μM) and displays minimal binding affinity for ghrelin and neurotensin-1 receptors (IC50s > 30 μM). Increases GLP-1 levels in vitro and in vivo. Reduces TNF-α-induced oxidative stress, mitochondrial dysfunction, cytokine expression and secretion of MMPs in fibroblast-like synoviocytes (FLS). Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | TC-G 1008 |
| CAS | 1621175-65-2 |
| Synonym | N-[3-Chloro-4-[[[2-(methylamino)-6-(2-pyridinyl)-4-pyrimidinyl]amino]methyl]phenyl]methanesulfonate |
| Quantity | 50 mg |
| Solubility | Soluble to 20mM in DMSO |
| Molecular Formula | C18H19ClN6O2S |
| Formula Weight | 418.9 |
| Physical Form | Solid |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
Research Use Only. Not For Human or Veterinary Use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

