Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TC-I 2014
GREENER_CHOICESDP

Catalog No. 541050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
541050 50 mg
541010 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 541050 Supplier Tocris Bioscience™ Supplier No. 5410/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent TRPM8 antagonist

TC-I 2014 is a potent TRPM8 antagonist (IC50 values are 0.8, 3.0 and 4.4 nM for canine, human and rat channels respectively). Exhibits antiallodynic properties in pain models in vivo.

Specifications

Chemical Name or Material TC-I 2014
CAS 1221349-53-6
Synonym 3-[7-(Trifluoromethyl)-5-[2-(trifluoromethyl)phenyl]-1H-benzimidazol-2-yl]-1-oxa-2-azaspiro[4.5]dec-2-ene
Quantity 50 mg
Solubility Soluble to 100mM in DMSO and to 100mM in ethanol
Molecular Formula C23H19F6N3O
Formula Weight 467.41
Physical Form Powder
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.