Learn More
R&D Systems™ Terminal Deoxynucleotidyl Transferase (TdT) Enzyme
SDP
Catalog No. 48103005
Change view
Click to view available options
Quantity:
30 μL
Catalog No. Quantity
48103005 30 μL
1 options
This item is not returnable. View return policy
Catalog No. 48103005 Supplier R&D Systems™ Supplier No. 48103005
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Suitable for use with TdT apoptosis or DNA damage and repair kits
Specifications
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 30 μL
Product Type Terminal Deoxynucleotidyl Transferase (TdT) Enzyme