Learn More
R&D Systems™ Universal Kinase Activity Kit, 2 Plate
Catalog No. ea004
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy
Includes
CHO cell-expressed Recombinant Mouse CD39L2/ENTPD6, ATP, ADP, Phosphatase Buffer 4, Phosphate Standard, Malachite Green Reagents A and B
A simple, nonradioactive high throughput-compatible format for assaying the enzyme activity of kinases in vitro
The majority of kinases use ATP as the phosphate donor. They transfer the terminal phosphate group of ATP to a substrate, producing ADP as a by-product.
Kit is an ADP-based phosphatase-coupled kinase assay that ues CD39L2/ENTPD6 as a coupling phosphatase
CD39L2 releases the beta-phosphate from ADP, and the released inorganic phosphate is detected using malachite green reagents
The amount of inorganic ph
Specifications
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 1 Kit
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.