Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ VU 0469650 Hydrochloride
GREENER_CHOICESDP

Catalog No. 537950 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
537950 50 mg
537910 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 537950 Supplier Tocris Bioscience™ Supplier No. 5379/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent and selective negative allosteric modulator of mGlu1

VU 0469650 hydrochloride is a potent and selective negative allosteric modulator of mGlu1 (IC50 = 99 nM). Exhibits >100-fold selectivity for mGlu1 over mGlu2-8 and 68 other GPCRs, ion channels, kinases and transporters. Brain penetrant.

Specifications

Chemical Name or Material VU 0469650 hydrochloride
CAS 1443748-47-7
Synonym 3-[(3R)-3-Methyl-4-(tricyclo[3.3.1.13,7]dec-1-ylcarbonyl)-1-piperazinyl]-2-pyridinecarbonitrile hydrochloride
Quantity 50 mg
Solubility Soluble to 100mM in DMSO and to 50mM in ethanol
Molecular Formula C22H28N4O·HCl
Formula Weight 400.94
Physical Form Solid
Product Type Bioactive Small Molecule
Purity >98%

Research Use Only. Not For Human or Veterinary Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.