Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (R)-DRF053 dihydrochloride
GREENER_CHOICE

Catalog No. 361010R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-101-0R 10 mg
36-105-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 36-101-0R Supplier Tocris Bioscience™ Supplier No. 3610/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent CK1 inhibitor; also inhibits cyclin-dependent kinases

(R)-DRF053 dihydrochloride is a potent casein kinase 1 (CK1) inhibitor (IC50 = 14 nM). Also an ATP-competitive cyclin-dependent kinase (cdk) inhibitor (IC50 values are 0.093 - 0.29, 0.21 and 0.82 μM for cdk2, cdk5 and cdk7, respectively). Shown to inhibit amyloid-β production in N2A-APP695 cells.

Specifications

Quantity 10 mg
Product Type (R)-DRF053 dihydrochloride
Purity >98%
Molecular Weight (g/mol) 490.43
Inhibitors (R)-DRF053 dihydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.