Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (R)-(+)-Etomoxir sodium salt
GREENER_CHOICESDP

Catalog No. 453950 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-395-0 50 mg
45-391-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-395-0 Supplier Tocris Bioscience™ Supplier No. 4539/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Carnitine palmitoyltransferase I (CPT1) inhibitor

Inhibitor of carnitine palmitoyltransferase I (CPT1); inhibits β-oxidation in mitochondria. Shown to inhibit cardiolipin biosynthesis from exogenous fatty acid in H9c2 cells.

Chemical Identifiers

CAS 828934-41-4
Molecular Formula C15H19ClNaO4

Specifications

Quantity 50 mg
Formula Weight 320.74
Purity >98%
Molecular Weight (g/mol) 321.75
Chemical Name or Material (R)-(+)-Etomoxir sodium salt
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.