Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-Glycidyl 3-Nitrobenzenesulfonate 98.0+%, TCI America™
SDP

Catalog No. G028625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G028625G 25 g
G02865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G028625G Supplier TCI America Supplier No. G028625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 115314-17-5
Molecular Formula C9H9NO6S
Molecular Weight (g/mol) 259.232
MDL Number MFCD00013436
InChI Key AIHIHVZYAAMDPM-MRVPVSSYSA-N
Synonym r---glycidyl nosylate, r-glycidyl nosylate, r-oxiran-2-ylmethyl 3-nitrobenzenesulfonate, r-glycidyl 3-nitrobenzenesulfonate, r---glycidyl 3-nitrobenzenesulfonate, r---glycidyl-3-nitrobenzenesulfonate, ccris 6393, oxiranylmethyl r-3-nitrobenzenesulfonate, benzenesulfonic acid, 3-nitro-, oxiranylmethyl ester, r
PubChem CID 146513
IUPAC Name [(2R)-oxiran-2-yl]methyl 3-nitrobenzenesulfonate
SMILES C1C(O1)COS(=O)(=O)C2=CC=CC(=C2)[N+](=O)[O-]

Specifications

Melting Point 63°C
Color White-Yellow
Quantity 25 g
UN Number 1325
Formula Weight 259.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-Glycidyl 3-Nitrobenzenesulfonate
RTECSNumber DB7192000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.