Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-Glycidyl Methyl Ether 97.0+%, TCI America™
SDP

Catalog No. G028025G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 64491-70-9
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00273372
InChI Key LKMJVFRMDSNFRT-BYPYZUCNSA-N
Synonym (R)-1-Methoxy-2,3-epoxypropane, (R)-2-(Methoxymethyl)oxirane
PubChem CID 2734441
IUPAC Name (2R)-2-(methoxymethyl)oxirane
SMILES COCC1CO1

Specifications

Color Yellow
Quantity 25 g
UN Number 1992
Formula Weight 88.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-Glycidyl Methyl Ether
EINECSNumber (2)-2425
RTECSNumber TZ3530000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.