Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human Glypican 3 Quantikine ELISA Kit
Click to view available options
Quantity:
1 Kit
Description
The Human Glypican 3 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human Glypican 3 in cell culture supernates (100 ul), cell lysates (20 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul).
Specifications
Specifications
Assay Range | 78.10 - 5000pg/mL |
Assay Sensitivity | 20.6pg/mL |
Detectable Analytes | Glypican 3 |
Assay | Solid Phase Sandwich ELISA |
Conjugate | HRP |
Cross Reactivity | < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules. |
Immunoassay Kit Format | 96-well Strip Plate |
Sample Type | Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum |
Specificity | Natural and Recombinant Human Glypican 3 |
For Use With (Application) | ELISA |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction