Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Klotho beta DuoSet ELISA

Catalog No. dy588905
Click to view available options
Quantity:
1 Kit (for 5 Plates)
This item is not returnable. View return policy
This item is not returnable. View return policy

Sufficient for at least five 96-well plates

The Human Klotho beta DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Klotho beta.

Specifications

Assay Range 31.20 to 2000 pg/mL
Detectable Analytes Klotho beta
Assay ELISA Development Kit
Conjugate Biotin
Cross Reactivity < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well microplate (sold separately)
Product Type Solid Phase Sandwich ELISA
For Use With (Application) ELISA
Gene ID (Entrez) 152831
Gene Symbol KLB
Host Species Human
Measuring Range 31.2-2,000pg/mL
Quantity 1 Kit (for 5 Plates)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym beta-klotho, betaKlotho, BKL, KLB, klotho beta, klotho beta like, klotho beta-like protein, MGC142213
Target Klotho beta
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.