Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Compass Biomedical™ R&D PLUS™ Human Platelet Lysate
SDP

Catalog No. PLS2
Encompass
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PLS2 250 mL
PLS1 100 mL
PLSA500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PLS2 Supplier Compass Biomedical™ Supplier No. PLS2
Add to Cart
Add to Cart

Advanced human platelet lysate that is safe and consistent for the rapid expansion of human mesenchymal cell cultures.

  • Potent: Human derived product rich in growth factors, cytokine, and other proteins
  • Products are growth factor rich supplements made from large pools of transfusion grade human platelets sourced from FDA-registered/AABB accredited blood centers.

Specifications

For Use With (Application) Cell Culture, Stem Cell Culture
Quantity 250 mL
Host Species Human
Product Type Human Platelet Lysate
Content And Storage Store at -80°C upon receipt. Thaw at room temperature or in a water bath. After thawing, aliquot and store at -20°C or colder. Long term storage at -80°C is recommended. Multiple freeze/thaw cycles are not recommended.
Format Bottle
Research Category RD
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.