Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-Limonene, ≥99% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111018030 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mL
5 mL
25 mL

(R)-(+)-Limonene is a terpenoid compound that is abundantly present in many plants such as rosemary. It is highly associated with plant and insect interactions due to its tendency to attract insects.

Featured Industries: Agriculture, Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 5989-27-5
Vapor Pressure <3 mmHg (14.4°C)
Vapor Density 4.7 (vs. air)
Density 0.842 g/mL (at 20°C)
Flash Point 51°C
Physical Form Neat
Linear Formula C10H16
MDL Number MFCD00062991
Boiling Point 176°C to 177°C
Synonym (+)-p-Mentha-1,8-diene; (+)-Carvene; (R)-4-Isopropenyl-1-methyl-1-cyclohexene
Type Analytical Standard
Quantity 25 mL
Formula Weight 136.23
Percent Purity ≥99% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.