Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Mouse Artemin DuoSet ELISA
Click to view available options
Quantity:
5 Plates
Description
The Mouse Artemin DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse Artemin.
Specifications
Specifications
Assay Sensitivity | 0pg/mL |
Detectable Analytes | Artemin |
Assay | ELISA Development Kit |
Conjugate | Biotin |
Cross Reactivity | < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules. |
Immunoassay Kit Format | 96-well microplate (sold separately) |
For Use With (Application) | ELISA |
Gene ID (Entrez) | 11876 |
Host Species | Mouse |
Measuring Range | 31.2-2,000pg/mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction