Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-N-(alpha-Methylbenzyl)phthalamic Acid 98.0+%, TCI America™
SDP

Catalog No. M16221G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 21752-35-2
Molecular Formula C16H15NO3
Molecular Weight (g/mol) 269.30
MDL Number MFCD00063050
InChI Key VCFKXWGKKDZMPO-LLVKDONJSA-N
PubChem CID 89038
IUPAC Name 2-{[(1R)-1-phenylethyl]carbamoyl}benzoic acid
SMILES C[C@@H](NC(=O)C1=CC=CC=C1C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 128°C
Color White
Quantity 1 g
Synonym r-2-1-phenylethyl carbamoyl benzoic acid, r-+-n-1-phenylethyl phthalamic acid, 2-1r-1-phenylethyl carbamoyl benzoic acid, r-+-n-alpha-methylbenzyl phthalamic acid, r-o-1-phenylethyl amino carbonyl benzoic acid, benzoic acid,2-1r-1-phenylethyl amino carbonyl, benzoic acid, 2-1r-1-phenylethyl amino carbonyl, +-n-1-phenylethyl phthalic acid mon, r-+-alpha-methylbenzyl phthalamic acid
Formula Weight 269.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-N-(alpha-Methylbenzyl)phthalamic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.