Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-N-BOC-(3-Pyridyl)alanine, 95%, 98% e.e., Thermo Scientific™

Catalog No. AC362930010
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98266-33-2
Molecular Formula C13H18N2O4
Molecular Weight (g/mol) 266.297
MDL Number MFCD00076901
InChI Key JLBCSWWZSSVXRQ-SNVBAGLBSA-N
Synonym boc-d-3-pyridylalanine, boc-d-3-pal-oh, boc-3-3-pyridyl-d-alanine, r-2-tert-butoxycarbonyl amino-3-pyridin-3-yl propanoic acid, r-n-boc-3-pyridyl alanine, boc-d-3-3-pyridyl-alanine, n-tert-butoxycarbonyl-3-pyridyl-d-alanine, boc-3'-pyridyl-d-ala, boc-3-3-pyridyl-d-ala-oh, boc-3-3'-pyridyl-d-alanine
PubChem CID 2734500
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-3-ylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CN=CC=C1)C(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Quantity 1 g
Formula Weight 266.29
Percent Purity ≥94%
Physical Form Powder
Chemical Name or Material (R)-N-BOC-(3-Pyridyl)alanine, 95%, (98% e.e.)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.