Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-p-Toluenesulfinamide 98.0+%, TCI America™
SDP

Catalog No. T28141G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28141G 1 g
T2814200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28141G Supplier TCI America Supplier No. T28141G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 247089-85-6
Molecular Formula C7H9NOS
Molecular Weight (g/mol) 155.215
MDL Number MFCD06858374
InChI Key YNJDSRPIGAUCEE-SNVBAGLBSA-N
PubChem CID 11051888
IUPAC Name 4-methylbenzenesulfinamide
SMILES CC1=CC=C(C=C1)S(=O)N

Specifications

Melting Point 118°C
Color White-Yellow
Quantity 1 g
Formula Weight 155.22
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(-)-p-Toluenesulfinamide
EINECSNumber (3)-1869
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.