Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ ProDots Recombinant Human TGF-beta 1
Click to view available options
Quantity:
250 μg
50 μg
Description
Includes
Individual vials include one ball of ProDots protein containing 5μg of Recombinant Human TGF-beta 1.
The ProDots Recombinant Human TGF-beta 1 Protein is derived from CHO. The ProDots Recombinant Human TGF-beta 1 Protein has been validated for the following applications: Bioactivity.
Order Info
Add ProDots proteins directly to the medium or reconstitute in the vial at 20μg/mL in 4mM HCl.
Specifications
Specifications
Accession Number | P01137 |
For Use With (Application) | Bioactivity |
Molecular Weight (g/mol) | Predicted MW: 12.8kDa (monomer) Observed MW: 12kDa, reducing conditions, 24kDa, non-reducing conditions |
Name | ProDots™ Recombinant Human TGF-beta 1 |
Quantity | 50 μg |
Source | Chinese Hamster Ovary cell line,CHO-derived human TGF-beta 1 protein Ala279-Ser390 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2-8° C as supplied. 1 month at 2-8° C under sterile conditions after reconstitution. 3 months at -20 to -80° C under sterile conditions after reconstitution. |
Structural Form | Disulfide-linked homodimer |
Gene Alias | CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 |
Conjugate | Unconjugated |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction