Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-Propylene Carbonate 98.0+%, TCI America™
SDP

Catalog No. P148525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P148525G 25 g
P14855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P148525G Supplier TCI America Supplier No. P148525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16606-55-6
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
MDL Number MFCD00798265
InChI Key RUOJZAUFBMNUDX-GSVOUGTGSA-N
Synonym (R)-4-Methyl-1,3-dioxolan-2-one
PubChem CID 641813
IUPAC Name (4R)-4-methyl-1,3-dioxolan-2-one
SMILES CC1COC(=O)O1

Specifications

Color Yellow
Quantity 25 g
Formula Weight 102.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-Propylene Carbonate
EINECSNumber (5)-0524
RTECSNumber FF9650000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.