Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-Propylene oxide, 98+%

Catalog No. p-4089820
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 15448-47-2 | C3H6O | 58.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15448-47-2
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00066211
InChI Key GOOHAUXETOMSMM-GSVOUGTGSA-N
Synonym r-+-propylene oxide, r-propylene oxide, 2r-2-methyloxirane, r-2-methyloxirane, r-methyloxirane, +-propylene oxide, r-epoxypropane, +-methyloxirane, r-+-1,2-epoxypropane, r-1,2-epoxypropane
PubChem CID 146261
ChEBI CHEBI:28985
IUPAC Name (2R)-2-methyloxirane
SMILES CC1CO1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -112.0°C
Density 0.8290g/mL
Boiling Point 33.0°C to 34.0°C
Flash Point −37°C
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 0.829
Solubility Information Solubility in water: 405g/L (20°C)
Viscosity 0.38 mPa.s (10°C)
Formula Weight 58.08
Percent Purity 98+%
Chemical Name or Material (R)-(+)-Propylene oxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled.
Harmful if swallowed.
May cause cancer.
Extremely flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic in contact with skin.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.