Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Quantikine Immunoassay Cntl Set 1027 Human CHI3L1/YKL-40
Click to view available options
Quantity:
1 Each
Specifications
Specifications
Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Detectable Analytes | Chitinase 3-like 1 |
Gene ID (Entrez) | 1116 |
Intended Use | Control |
Quantity | 1 Each |
Source | Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction