Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Quantikine Immunoassay Control Set 848 for Human Glypican 3

Catalog No. QC203
Click to view available options
Quantity:
1 Each
This item is not returnable. View return policy
This item is not returnable. View return policy

For use with catalog number DGLY30

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes Glypican 3
Gene ID (Entrez) 2719
Intended Use Control
Quantity 1 Each
Source Human
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.