Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R,R)-2,2'-Isopropylidenebis(4-phenyl-2-oxazoline) 96.0+%, TCI America™
SDP

Catalog No. D28231G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28231G 1 g
D2823250MG 250 mg
D28235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D28231G Supplier TCI America Supplier No. D28231G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 150529-93-4
Molecular Formula C21H22N2O2
Molecular Weight (g/mol) 334.42
MDL Number MFCD00192244
InChI Key JTNVCJCSECAMLD-UHFFFAOYNA-N
Synonym +-2,2'-isopropylidenebis 4r-4-phenyl-2-oxazoline, ph-box, r,r, unii-cg7p67w7w0, r,r-2,2'-isopropylidenebis 4-phenyl-2-oxazoline, r,r-2,2'-dimethylmethylene bis 4-phenyl-2-oxazoline, 4r,4'r-2,2'-propane-2,2-diyl bis 4-phenyl-4,5-dihydrooxazole, oxazole, 2,2'-1-methylethylidene bis 4,5-dihydro-4-phenyl-, 4r,4'r, r,r-2,2-bis 4-phenyl-2-oxazolin-2-yl propane, r,r-2,2-dimethylmethylene bis 4-phenyl-2-oxazoline
PubChem CID 676390
IUPAC Name 4-phenyl-2-[2-(4-phenyl-4,5-dihydro-1,3-oxazol-2-yl)propan-2-yl]-4,5-dihydro-1,3-oxazole
SMILES CC(C)(C1=NC(CO1)C1=CC=CC=C1)C1=NC(CO1)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 334.42
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (R,R)-2,2′-Isopropylidenebis(4-phenyl-2-oxazoline)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.