Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R,R)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine 98.0+%, TCI America™
SDP

Catalog No. B22171G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22171G 1 g
B2217250MG 250 mg
B22175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B22171G Supplier TCI America Supplier No. B22171G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 131864-67-0
Molecular Formula C17H23N3O2
Molecular Weight (g/mol) 301.39
MDL Number MFCD01321334
InChI Key CSGQGLBCAHGJDR-GJZGRUSLSA-N
PubChem CID 688213
IUPAC Name (4R)-4-propan-2-yl-2-[6-[(4R)-4-propan-2-yl-4,5-dihydro-1,3-oxazol-2-yl]pyridin-2-yl]-4,5-dihydro-1,3-oxazole
SMILES CC(C)C1COC(=N1)C2=NC(=CC=C2)C3=NC(CO3)C(C)C

Specifications

Melting Point 154°C
Color White-Yellow
Quantity 1 g
Synonym (R,R)-2,2′C-(2,6-Pyridinediyl)bis(4-isopropyl-2-oxazoline)
Formula Weight 301.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R,R)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.