Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Rabbit IL-4 DuoSet ELISA

Catalog No. DY6939
Click to view available options
Quantity:
15 Plate
This item is not returnable. View return policy
This item is not returnable. View return policy

Rabbit IL-4 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Rabbit IL-4 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies rabbit IL-4.

Specifications

Assay Range 4.69 - 300pg/mL
Assay Sensitivity 0pg/mL
Detectable Analytes IL-4
Assay ELISA Development Kit
Conjugate Biotin
Cross Reactivity < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well microplate (sold separately)
For Use With (Application) ELISA
Gene ID (Entrez) 100302454
Host Species Rabbit
Measuring Range 4.69-300pg/mL
Quantity 15 Plate
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.