Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Yeast GST-Ubiquitin Activating Enzyme (UBE1)

Catalog No. E300050
Click to view available options
Quantity:
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Recombinant Yeast GST-Ubiquitin E1 Enzyme (UBE1) is a member of the Ubiquitin-activating (E1) enzyme family that is required for the first step of the enzymatic cascade that subsequently utilizes a Ubiquitin-conjugating (E2) enzyme and a Ubiquitin ligase (E3) to conjugate Ubiquitin to substrate proteins. Reaction conditions will need to be optimized for each specific application. We recommend an initial GST-Ubiquitin E1 Enzyme (UBE1) concentration of 50-200 nM.

Order Info

The product is shipped with dry ice or equivalent. Upon receipt, store it immediately

Specifications

Accession Number P22515
For Use With (Application) Bioactivity
Formulation 0.71 mg/mL (5 μM) in 50 mM HEPES pH 8.0, 100 mM NaCl, 10% Glycerol (v/v), 1 mM TCEP
Gene ID (Entrez) 7317
Molecular Weight (g/mol) M.W. (theoretical): 141 kDa
Name GST Ubiquitin-activating Enzyme/UBE1, Carrier Free
Quantity 50 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived yeast Ubiquitin-activating Enzyme/UBE1 protein Contains an N-terminal GST (glutathione S-transferase) tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Symbol UBA1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein Ubiquitin-activating Enzyme/UBE1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.