Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (R,S)-Bode Kinetic Resolution Catalyst

Catalog No. 804568100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-456-8100MG 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 80-456-8100MG Supplier Sigma Aldrich Supplier No. 804568100MG
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The following hydroxamic acid catalyst was shown by Bode and coworkers to assist in the kinetic resolution of cyclic amines through acylation of one enantiomer, which can be controlled using either the (R or S)- or (S or R)-Bode Kinetic Resolution Catalyst, both of which are also available through Sigma-Aldrich. It can also be employed in the preparation of nonracemic piperidines by kinetic resolution using an +a-hydroxy-+a'sa, +a-unsaturated ketone in the presence of a pyrrolotriazolium salt.Expanded substrate scope and catalyst optimization for the catalytic kinetic resolution of N-heterocycles

Specifications

Melting Point 138°C to 140°C
Molecular Formula C37H67NO13 -+ xH2O
Linear Formula C37H67NO13 -+ xH2O
MDL Number MFCD00149235
Quantity 5 g
Molecular Weight (g/mol) 733.93 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.