Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R-Spondin 2 Mouse anti-Human, Clone: 14C7.1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS1709
Description
Anti-R-Spondin 2, clone 14C7.1 is a mouse monoclonal antibody that specifically targets R-Spondin 2 (RSPO2) and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Specifications
R-Spondin 2 | |
Monoclonal | |
Please refer to lot specific datasheet. | |
RSPO2,UNQ9384/PRO34209 | |
GST/His-tagged recombinant fragment corresponding to 74 amino acids from the C-terminal end of human R-Spondin 2. | |
100 μg | |
Signaling | |
NP_848660 | |
Human | |
Purified |
Immunohistochemistry (Paraffin), Western Blot | |
14C7.1 | |
Unconjugated | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Clone 14C7.1 specifically detects human R-spondin-2. It targets an epitope within 74 amino acids from the C-terminal end. | |
Stable for 1 year at 2-8°C from date of receipt. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction