Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R-Spondin 2 Mouse anti-Human, Unconjugated, Clone: 14C7.1, MilliporeSigma™

Catalog No. MABS170925 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1709-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1709-25 Supplier MilliporeSigma Supplier No. MABS170925UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-R-Spondin 2, clone 14C7.1, Cat. No. MABS1709, is a mouse monoclonal antibody that specifically targets R-Spondin 2 (RSPO2) and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen R-Spondin 2
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 14C7.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Roof plate-specific spondin-2;hRspo2
Gene Symbols RSPO2;UNQ9384/PRO34209
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 74 amino acids from the C-terminal end of human R-Spondin 2.
Molecular Weight of Antigen ∽35 kDa observed; 28.31 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone 14C7.1 specifically detects human R-spondin-2. It targets an epitope within 74 amino acids from the C-terminal end.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.