Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R-Spondin 3 Antibody (OTI3D5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245637
Description
Ensure accurate, reproducible results in Immunohistochemistry
R-Spondin 3 Monoclonal specifically detects R-Spondin 3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
R-Spondin 3 | |
Monoclonal | |
1 mg/mL | |
Immunohistochemistry 1:150 | |
CRISTIN1, FLJ14440, hPWTSR, hRspo3, Protein with TSP type-1 repeat, PWTSR, Roof plate-specific spondin-3, R-spondin 3 homolog (Xenopus laevis), R-spondin-3, Thrombospondin type-1 domain-containing protein 2, thrombospondin, type I, domain containing 2, THSD2 | |
Mouse | |
30.7 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Immunohistochemistry | |
OTI3D5 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
RSPO3 | |
Human recombinant protein fragment corresponding to amino acids 22-272 of human RSPO3 (NP_116173) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
84870 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction