Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer solution pH 7, Phosphate buffer

Catalog No. AC258590025
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L

CAS: 7558-79-4 | H2KO4P

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

pH buffer
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer solution pH 7
CAS 7558-79-4, 7732-18-5
Density 1.0030g/mL
Quantity 2.5 L
Molecular Formula H2KO4P
MDL Number MFCD03097944
Packaging Plastic bottle
Specific Gravity 1.003
Merck Index 14, 7659
Vapor Pressure 14mmHg at 20°C
Solubility Information Solubility in water: soluble.
Formula Weight 136.09
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.